Would you like to log in again?
Nexus™ Horizontal Clean Bench, 4' Wide
Supplier: Labconco
Nexus™
5340600 5340310 5341600 5340800 5340210 5341810 5341610 5341500 5341700 5341210 5341000 5340810 5340300 5340110 5340000 5340610 5340010 5341310 5341110 5341300 5340500 5341200 5341510 5341800 5340510 5341010 5341100 5340100 5340710 5341710 5340200 5340700
77037-848EA 11220 USD InStock
77037-848 77037-804 77037-826 77037-802 77037-846 77037-824 77037-808 77037-828 77037-806 77037-850 77037-810 77037-854 77037-832 77037-852 77037-830 77037-814 77037-836 77037-858 77037-856 77037-812 77037-834 77037-818 77037-838 77037-816 77037-862 77037-840 77037-860 77037-800 77037-822 77037-844 77037-820 77037-842
Nexus™ Horizontal Clean Bench, 4' Wide
Fume Hoods and Enclosures Laminar Flow Cabinets
Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area.
  • Efficient unidirectional laminar airflow provided through CAP™ technology
  • Intelligent 5" touchscreen interface features intuitive monitoring capabilities
  • Reliable with long-life HEPA filters and maximum cleanability that stands up to disinfectants
The Nexus horizontal clean bench is a laminar flow workstation designed to protect non-hazardous samples and processes from contamination. The Nexus provides an ISO 4 clean air environment.
The Nexus is the most advanced clean bench on the market. New features include a touchscreen interface, Constant Airflow Profile (CAP™) technology that maintains proper airflow velocity as the HEPA filter loads, the ability to log all operational parameters for compliance with regulatory requirements, clear and detailed alarm and alert notifications, and a full OS with user-customizable features, maintenance logs and diagnostics.
Certifications: The 230 V units are CE/UKCA Certified.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty