Would you like to log in again?
Nexus™ Horizontal Clean Bench, 5' Wide
Supplier: Labconco
Nexus™
5350610 5350500 5350300 5351000 5350200 5351800 5350110 5350310 5351810 5350800 5351100 5351710 5351010 5350010 5351610 5351700 5351110 5351300 5350000 5351210 5350510 5350810 5351200 5350600 5350210 5350710 5350100 5351500 5351510 5351600 5350700 5351310
77037-924EA 14660 USD InStock
77037-924 77037-902 77037-868 77037-900 77037-922 77037-906 77037-926 77037-904 77037-908 77037-890 77037-872 77037-894 77037-870 77037-892 77037-910 77037-876 77037-898 77037-896 77037-874 77037-914 77037-912 77037-878 77037-918 77037-916 77037-880 77037-884 77037-882 77037-866 77037-888 77037-920 77037-886 77037-864
Nexus™ Horizontal Clean Bench, 5' Wide
Fume Hoods and Enclosures Laminar Flow Cabinets
Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area.
  • Efficient unidirectional laminar airflow provided through CAP™ technology
  • Intelligent 5" touchscreen interface features intuitive monitoring capabilities
  • Reliable with long-life HEPA filters and maximum cleanability that stands up to disinfectants
The Nexus Horizontal clean bench is a laminar flow workstation designed to protect non-hazardous samples and processes from contamination. The Nexus provides an ISO 4 clean air environment.
The Nexus is the most advanced clean bench on the market. New features include a touchscreen interface, Constant Airflow Profile (CAP™) technology that maintains proper airflow velocity as the HEPA filter loads, the ability to log all operational parameters for compliance with regulatory requirements, clear and detailed alarm and alert notifications, and a full OS with user-customizable features, maintenance logs and diagnostics.
Certifications: The 230 V units are CE/UKCA Certified.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty