Would you like to log in again?
Nexus™ Horizontal Clean Bench, 6' Wide
Supplier: Labconco
Nexus™
5361610 5361600 5361100 5360010 5361310 5360810 5360500 5361700 5360000 5361110 5361710 5361000 5361200 5360110 5360600 5361210 5360510 5360200 5361810 5360700 5360300 5361500 5360100 5361300 5361510 5360800 5361800 5360310 5361010 5360710 5360210 5360610
77037-968EA 14980 USD InStock
77037-968 77037-946 77037-944 77037-966 77037-988 77037-928 77037-948 77037-990 77037-972 77037-950 77037-970 77037-976 77037-932 77037-954 77037-930 77037-974 77037-952 77037-958 77037-936 77037-934 77037-978 77037-956 77037-938 77037-982 77037-960 77037-980 77037-964 77037-986 77037-942 77037-984 77037-940 77037-962
Nexus™ Horizontal Clean Bench, 6' Wide
Fume Hoods and Enclosures Laminar Flow Cabinets
Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area.
  • Efficient unidirectional laminar airflow provided through CAP™ technology
  • Intelligent 5" touchscreen interface features intuitive monitoring capabilities
  • Reliable with long-life HEPA filters and maximum cleanability that stands up to disinfectants
The Nexus Horizontal clean bench is a laminar flow workstation designed to protect non-hazardous samples and processes from contamination.
The Nexus is the most advanced clean bench on the market. New features include a touchscreen interface, Constant Airflow Profile (CAP™) technology that maintains proper airflow velocity as the HEPA filter loads, the ability to log all operational parameters for compliance with regulatory requirements, clear and detailed alarm and alert notifications, and a full OS with user-customizable features, maintenance logs and diagnostics.
Certifications: The 230 V units are CE/UKCA certified.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty