Would you like to log in again?
Nexus™ Horizontal Clean Bench, 8' Wide
Supplier: Labconco
Nexus™
5380610 5380010 5381100 5381810 5380300 5381310 5380600 5381600 5380500 5380110 5380710 5380000 5381710 5380310 5381800 5380100 5381300 5381200 5381210 5381010 5381510 5381700 5380200 5381110 5380810 5381610 5380510 5380800 5380210 5381000 5380700 5381500
77038-050EA 18690 USD InStock
77038-050 77038-052 77038-030 77038-054 77038-032 77038-010 77038-012 77038-034 77038-014 77038-036 77038-038 77038-016 77038-018 77038-008 77037-994 77037-992 77037-998 77037-996 77038-040 77038-042 77038-020 77038-044 77038-000 77038-022 77038-046 77038-002 77038-024 77038-004 77038-026 77038-048 77038-028 77038-006
Nexus™ Horizontal Clean Bench, 8' Wide
Fume Hoods and Enclosures Laminar Flow Cabinets
Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area.
  • Efficient unidirectional laminar airflow provided through CAP™ technology
  • Intelligent 5" touchscreen interface features intuitive monitoring capabilities
  • Reliable with long-life HEPA filters and maximum cleanability that stands up to disinfectants
The Nexus Horizontal clean bench is a laminar flow workstation designed to protect non-hazardous samples and processes from contamination.
The Nexus is the most advanced clean bench on the market. New features include a touchscreen interface, Constant Airflow Profile (CAP™) technology that maintains proper airflow velocity as the HEPA filter loads, the ability to log all operational parameters for compliance with regulatory requirements, clear and detailed alarm and alert notifications, and a full OS with user-customizable features, maintenance logs and diagnostics.
Certifications: The 230 V units are CE/UKCA certified.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty