Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Labconco® Heated Product Shelf Chambers with Valves

Supplier: Labconco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

780802030 780802100 780802015 780802190 780802040 780802150 780802070 780802130 780802170 780802110 780802090 780802010 780802000 780802140 780802115 780802050
77147-854EA 14795.55 USD InStock
77147-854 77147-844 77147-866 77147-852 77147-842 77147-864 77147-850 77147-840 77147-862 77147-860 77147-838 77147-858 77147-848 77147-856 77147-868 77147-846
Labconco® Heated Product Shelf Chambers with Valves
Furniture Shelves
Heated product shelf chambers with valves.

  • Includes 12" dia. acrylic cylinder and base, 10" dia. removable shelves, and temperature probes
  • Controlled heat settings−20 to 50 °C (−4 to 140 °F)
  • Shelves do not cool
  • Has 8" dia. opening and cap for 3" dia. attachment port

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty