Would you like to log in again?
SERVICE FIXTURE KIT-HORIZ. CB
Supplier: Labconco
Service Fixture Kit^ For Purifier* Horizontal Clean Bench. Includes serrated-hose-tip valve of chrome-plated forged-brass with quarter turn handle, hardware and instructions for plumbing to services. For field installation.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SERVICE FIXTURE KIT-HORIZ. CB 82000-982 3773700 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty