Would you like to log in again?
CHARCOAL FILTER 4FT
Supplier: Labconco
Replacement Activated Carbon with Alumina Exhaust Filter^ for 4' PuriCare Procedure Station. Granular activated carbon and impregnated alumina media adsorb odors, pollutants and light gases.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHARCOAL FILTER 4FT 82005-588 3796200 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty