Would you like to log in again?
Explosion-Proof Blowers for Explosion-Proof Laboratory Hoods, Labconco®
Supplier: Labconco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7069700 7069100 7180500 7180300 7182300 7068900 7180700 7068700 7181100 7069300 7181300 7181500 7180900 7180100 7181700 7068500 7068300 7182100 7069500 7068100
82006-770EA 4885 USD InStock
82006-770 82006-798 82006-764 82006-800 82006-766 82006-810 82006-761 82006-772 82006-774 82006-762 82006-806 82006-816 82006-808 82006-818 82006-802 82006-758 82006-768 82006-812 82006-804 82006-814
Explosion-Proof Blowers for Explosion-Proof Laboratory Hoods, Labconco®
Air Purifiers
Fiberglass explosion-proof blowers are designed for fume hood exhaust systems in moderate to highly corrosive conditions and have non-sparking polypropylene impellers and explosion-proof motors.
Coated steel explosion-proof blowers are recommended for low to moderately corrosive applications and feature non-sparking aluminum impellers and explosion-proof motors for maximum safety. Housing has phenolic coating for corrosion resistance.
Both blowers have a dry powder epoxy-coated base and weather cover for protection in extreme weather. Sheaves are adjustable to allow for air volume adjustment. Self-adjusting gravity belt tightener ensures consistent performance because belt is held taught by the weight of the motor. Motor mount keeps the motor in place should the belt ever fail.
Housing is molded fiberglass-reinforced polyester for durability and long life.
Certifications: Blower motors are UL listed.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty