Would you like to log in again?
FreeZone® Tube Holders for Freeze Dryers, Labconco®
Supplier: Labconco
FreeZone®
7379500 7442600 7379400 7442700
75841-030EA 378.94 USD InStock
75841-030 82010-522 75841-032 82010-520
FreeZone® Tube Holders for Freeze Dryers, Labconco®
Racks Tube Racks
Hold tubes in upright position within a flask for freeze drying.
  • Sturdy steel or aluminum construction
  • Designed to fit securely inside Fast-Freeze® and Lyph-Lock® Flasks
  • Accommodate a variety of tube sizes
Stainless steel tube holders keep test tubes in an upright position and fit inside flasks for easy transport and freeze drying
Holders for 9 each (15mL) 17 × 120 mm tubes and 4 each (50mL) 30 ×115mm tubes each fit inside a 900mL or larger flask.
Holders for 15 each 12 or 13mm tubes, and 30 each 2mL microcentrifuge tubes each fit inside a 750mL or larger flask.
Tubes and glassware not included.
Ordering information: Tubes and glassware are not included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty