Would you like to log in again?
Manual Duct Dampers, Labconco
Supplier: Labconco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4724200 4741300 5983400 4726400 5981200
82006-952EA 940 USD InStock
82006-952 82006-954 82010-726 82006-950 82006-948
Manual Duct Dampers, Labconco
Ducting Supplies
Damper fitting allows airflow balance
The duct dampers can be used with exhaust and auxiliary air ducts, and are usually placed directly above the fume hood.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty