Would you like to log in again?
XPert™ Filtered Balance Stations, Labconco®
Supplier: Labconco
XPert™
3946532 3940200 3940422 3946622 3946502 3940222 3946541 3940201 3946531 3946632 3940301 3946500 3940420 3946522 3940401 3940221 3946601 3946602 3940300 3946600 3940321 3940302 3940322 3946521 3940320 3946631 3946501 3946630 3946620 3946621 3946540 3946641 3946642 3940400 3946542 3946640 3946520 3940402 3940202 3946530 3940220 3940421
82018-136EA 8120 USD InStock
82018-136 82018-138 82018-154 82018-132 82018-134 93000-580 82018-140 93000-588 93000-566 93000-600 93000-564 82018-142 89004-500 93000-586 93000-584 89004-502 89004-504 93000-582 82018-148 93000-608 93000-606 82018-144 93000-604 93000-568 82018-146 93000-602 93000-592 93000-570 93000-590 89004-494 89004-496 89004-498 93000-610 93000-578 82018-150 93000-598 93000-576 82018-152 93000-574 93000-596 93000-572 93000-594
XPert™ Filtered Balance Stations, Labconco®
Fume Hoods and Enclosures Balances Enclosures
These stations provide user protection by keeping powders and particulates contained during weighing procedures
The deep interiors easily accommodate large micro and analytical balances. The Clean-Sweep™ airfoil allows air to sweep the work surface for maximum containment. The perforated rear baffle promotes horizontal laminar airflow to maximize containment. The upper containment sash foil bleeds air into the enclosure to direct concentrations of contaminants away from the operator’s breathing zone. The upper dilution air supply introduces room air at the top of the sash to dilute contaminants in the upper chamber and bathe the back of the sash with clean air. Additional side-entry airfoils allow air to sweep across the interior glass surfaces to enhance containment. The airfoil’s ergonomic design provides a comfortable position for the operator’s forearms.
Stations feature a true bag-in/bag-out filter disposal system for safely removing and replacing the HEPA filter. The HEPA filter cell is 99.99% efficient on particulates 0.3µm in size. An upper diffuser screen, located above the work area, provides protection for the HEPA filter. The station also features a front-mounted Minihelic® pressure gauge that monitors pressure differential across the HEPA filter to help determine filter loading.
Sturdy glacier white and gray, dry powder epoxy-coated aluminum frame and steel rear plenum and baffle dissipate static charge. Tempered safety glass front sash, sides, and top offer excellent visibility, ambient light, and protection. The ergonomic angled sash allows a closer view, reduces glare, and provides a more comfortable operating position than vertical sashes. The hinged sash pivots upward and locks to a loading height of 48.3cm (19") on the 0.6m (2'), 0.9m (3'), and 1.2m (4') models. The 1.5m (5') and 1.8m (6') models incorporate a gas-assist lift and have a loading height of 70.4cm (2711/16"). The operating sash height is 23.9cm (913/32") on the 0.6m (2'), 0.9m (3'), and 1.2m (4') models. The 1.5m (5') and 1.8m (6') models have a 30.5cm (12") operating sash height. A wiping seal helps to keep contaminants contained. Stations also include front-mounted light and blower switches, two utility ports, and glare-free fluorescent lighting.
Enclosures also feature an optional built-in airflow monitor. If the hood’s face velocity falls below the setpoint for more than six seconds, an audible alarm sounds and a red light glows. The alarm can be muted. On the Guardian™ monitor, an LED glows green when the airflow is within setpoint range. The Guardian 1000 digital monitor features an LCD screen of actual face velocity and an RS-232 port. Stainless steel work surfaces feature a spill trough. The utility shelf kit has three epoxy-coated steel shelves that can hold a printer, utensils, and other miscellaneous supplies. Shelves attach to the rear baffle perforations of the enclosure.
The built-in HEPA filter is mounted in the upper plenum and an exhaust collar allows connection to 15.2cm dia. (6") ductwork and a remote blower.
Certifications: All stations conform to modified ASHRAE 110-1995 standards, SEFA 1-2002 standards, and ANSI Z9.5-1-993 standards. The 115V models are ETL and ETL-C listed; the 230V models are CE marked.
Ordering information: All stations require a work surface, supporting base, remote blower, and ductwork. For base cabinets, see the Protector® acid storage cabinets (46610-412 series), solvent storage cabinets (46610-440 series), and standard cabinets (46610-404 series). The 1.5m (5') and 1.8m (6') models with an exterior depth of 93.5cm (3613/16") include a left side-mounted waste chute that accepts bags 25.4x40.6cm (10x16") and larger. The 230V models do not include a plug. Items designated below as REDISHIP can be shipped within 48 hours of receiving your order.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty