Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Purifier® Vertical Clean Benches, Labconco®
Supplier: Labconco
Purifier®
3970300 3970304 3970420 3970404 3970220 3970403 3970201 3970423 3970321 3970204 3970301 3970401 3970320 3970223 3970421 3970221 3970324 3970203 3970424 3970323 3970200 3970224 3970400 3970303
89000-070EA 12110 USD InStock
89000-070 89000-060 89000-072 89000-050 89000-062 89000-140 89000-052 89000-074 89000-138 89000-130 89000-064 89000-076 89000-142 89000-054 89000-132 89000-066 89000-144 89000-078 89000-056 89000-134 89000-068 89000-058 89000-136 89000-048
Purifier® Vertical Clean Benches, Labconco®
Fume Hoods and Enclosures Laminar Flow Cabinets
Clean benches provide product protection from environmental contaminants for applications requiring a particulate-free work area, including plant tissue culture, electronic part inspection, syringe filling, medical device assembly, media plate preparation, and PCR
Base stands (sold separately) are made of durable 4.4 cm (1³/₄") thick, tubular, glacier white, epoxy-coated steel. The height of base stands is fully adjustable from 69.9 to 85.1 cm (27¹/₂ to 33¹/₂") in 2.5 cm (1") increments. Base stands with fixed feet come with leveling feet for use on uneven floor surfaces. Base stands with casters include four 12.7 cm (5") non-marking casters. The accessory shelf for telescoping base stands hooks onto the lower rear horizontal cross member of the stand and provides space for miscellaneous accessories and apparatus. Hydraulic stand height is infinitely adjustable from 64.8 to 85.1 cm (25¹/₂ to 33¹/₂"). Height adjustments can be made any time on hydraulic stands. Caster kit adds mobility to the hydraulic lift base stand. Solid epoxy dished work surfaces (recommended for operation) are also available. IV bar supports intravenous solution bottles and bags.
The angled sash provides closer, more comfortable viewing than vertical sashes and has a working sash height of 24 cm (9⁷/₁₆"). It pivots upward and locks to a loading height of 50.8 cm (20"). The front-mounted blower and light switches are located within easy reach of the operator, and two utility ports allow passage of tubing and electrical cords through the back.
Variable speed blowers with solid-state speed control maintain proper air velocities. A diffuser, located at the top of the work area, stabilizes the airflow through the work area and protects the surface of the HEPA filter from possible damage from apparatus inside the enclosure, while a rear exhaust grill minimizes air turbulence in the work area. A prefilter traps large particles to extend the life of the HEPA filter, which removes 99.99% of all particles >=0.3 µm, and an aerodynamic airfoil allows filtered air to sweep the work surface, promoting ISO Class 5 (FED-STD-209E Class 100/M3.5) conditions. In addition, a front-mounted Minihelic® pressure gauge monitors differential pressure across the HEPA filter.
Tempered safety glass sash and side panels provide a naturally illuminated work area. A fluorescent light is located outside of the work space for easy access, and an optional UV light may be used in conjunction with surface disinfection to ensure thorough decontamination while the cabinet is unattended. On models with a UV light, a single three-way switch powers the fluorescent and UV lights, but only one can be on at a time. Models with UV light also have a variable digital timer that provides continuous or timed UV light exposure in intervals of 5, 10, 15, 30, 60, 120, and 240 minutes and a UV protection panel that allow the sash opening to be closed while the UV light is on and prevent contaminants from entering when the clean bench is not in use. An optional built-in Guardian™ airflow monitor continuously monitors airflow and alerts the user to velocities outside of the preset range through LED and alarm indicators.
Certifications: The 115 V enclosures are ETL and ETL-C listed with CAN/CSA C22.2 No. 1010.1 conformity. The 230 V models are CE marked.
Ordering information: Manufacturer’s one-year warranty on parts and labor. Base stand and work surface (recommended for operation) are not included. An interlocked UV Protection Panel replaced the bi-fold doors. Variable digital timer for the UV lamp is now a standard feature.
Order Now
ORDER
ACCESSORIES
SERVICES
Services: VWR CATALYST™ offers customer on-site validation, Testing & certification services including Face Velocity Profile Test, Smoke Generation Test, Tracer Gas Test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWRCATALYST@vwr.com for more details. You can also visit https://vwr.com/equip_services.
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty