Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Protector® Downdraft Powder Station, Labconco
Supplier: Labconco
3410021 3410025 3410023 3410024 3410022 3410000 3410001 3410020
89167-538EA 4575 USD InStock
89167-538 89167-536 89167-548 89167-546 89167-544 89167-542 89167-550 89167-540
Protector® Downdraft Powder Station, Labconco
Fume Hoods and Enclosures Forensic Enclosures
Protector® Downdraft Powder Stations keep users safe by containing powders during fingerprint processing. The open-sided, open-top work station provides unrestricted operator movement and accommodates oversized evidence. Available in 2' and 3' widths, two or more Stations may be placed side-by-side to achieve an unlimited number of width combinations. An optional Transition Connector mounts between two Stations for a secure fit and continuous work surface.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty