Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
CentriVap® micro IR Vacuum Concentrator 230 V
Supplier: Labconco
7701035 7701037 7701030
89177-054EA 10868.1 USD InStock
89177-054 89177-048 89177-046
CentriVap® micro IR Vacuum Concentrator 230 V
Evaporators Centrifugal Evaporators
With the smallest footprint available, the new CentriVap® micro IR Vacuum Concentrator fits in tight spaces and is ideal for small throughput in molecular biology, proteomics, genomics, genetics, cell biology and drug discovery labs
  • 230 V model
Everything about the CentriVap® micro IR is small except its many features. Its transparent infrared (IR) glass lid directs heat to the samples to speed evaporation and reduces cross contamination from condensation. The CentriVap® micro IR includes a rotor for twenty-four small samples, a CentriZap® micro IR strobe light to view samples while the rotor is spinning, an built-in diaphragm pump and glass trap everything you need to start concentrating. Simply plug it in and it's ready to use.
Weighing only 20 pounds, this personal-sized concentrator may be easily transported from lab to lab.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89177-044) is also available, please see separate product page.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty