Would you like to log in again?
4FT PRE-FILTER KIT LG
Supplier: Labconco
Prefilter Starter Kit For Use In 4' Purifier* Logic* And Purifier* Cell Logic* Biosafety Cabinets Includes towel catch; washable, screen-type prefilter and hardware including bracket for installing on the towel catch. installation is required. ea.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
4FT PRE-FILTER KIT LG 89203-028 3854801 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty