Labconco Protector® Premier® fiberglass fume hoods have a one-piece liner of specially-formulated, fiberglass-reinforced polyester, which offers corrosion and fire resistance and easy clean up.
These by-pass hoods feature the largest unobstructed sightline available (37.5" from work surface to header panel). Other features include Eco-Foil™ air foil that corrects natural turbulence while keeping cords out of the way, dry powder-coated steel exterior, safety glass sash, and molded fiberglass exhaust collar. Standard hoods include pre-wired LED lighting rated for 50000 hours. Explosion-proof (EP) models include incandescent lighting (bulb not included).
Hoods are available with a built-in standard fiberglass blower, or an explosion-proof fiberglass blower, or without a blower (remote blower sold separately).
There are no seams where deterioration can occur or where contaminants can collect. These general purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.
Certifications: All models are ETL listed. 230V models are CE listed.
Ordering information: Customize your hood with optional service fixtures that accommodate air, gas, vacuum or water lines. Complete your fume hood with a variety of accessories including blowers, work surfaces, storage cabinets, fire extinguishers, and air flow monitors (sold separately).