Would you like to log in again?
CentriVap® Complete™ Vacuum Concentrator, Labconco®
Supplier: Labconco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CentriVap®
7315022 7315060 7315063 7315042 7315072 7315033 7315040 7315031 7315023 7315020 7315043 7315062 7315073 7315030 7315071 7315070 7315032 7315041 7315021 7315061
89259-752EA 30756.97 USD InStock
89259-752 89259-750 89239-660 89239-650 89239-672 89239-670 89239-664 89239-642 89239-654 89239-640 89239-662 89239-652 89239-668 89239-646 89239-658 89239-644 89239-666 89239-656 89239-648 89239-638
CentriVap® Complete™ Vacuum Concentrator, Labconco®
Concentrators
The all-in-one CentriVap® Complete Vacuum Concentrator houses every component needed to begin processing multiple samples
Using centrifugal force, vacuum and controlled heat, the CentriVap® Complete speeds concentration of samples in a variety of applications such as molecular biology, proteomics, genomics, cell biology, microbiology, biochemistry, drug discovery, and analytical chemistry. Acid-resistant models are available for corrosive chemical use. Other models feature Heat Boost™, which surrounds the chamber wall to further speed the evaporation of high boiling point solvents.
The vacuum concentrator, -50°C cold trap, diaphragm pump, rotor, and CentriZap™ strobe light are all included. Simply add samples in the containers of your choice and start concentrating.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty