Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VACUUBRAND® Aspirator Pumps, Labconco
Supplier: Labconco
3850332 3850330 3850310 3850333 3850340 3850335 3850345 3850341 3850331 3850320 3850342 3850343 3850344 3850334
89410-188EA 2020 USD InStock
89410-188 89410-210 89410-200 89410-198 89410-196 89410-194 89410-208 89410-206 89410-214 89410-204 89410-212 89410-202 89410-192 89410-190
VACUUBRAND® Aspirator Pumps, Labconco
Pumps Vacuum Aspiration Systems
BVC Basic Liquid Aspiration Systems are designed for use with existing vacuum supplies, individual diaphragm pumps and house vacuum systems
Both systems are convenient to cell culture work or other laboratory applications, are compact and offer a flexible, integrated solution for filtration and efficient aspiration of supernatants. The units are compatible with polypropylene or borosilicate glass vacuum bottles as indicated.
BVC Control Liquid Aspiration Systems are for use in labs without in-house vacuum.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty