Would you like to log in again?
CENTRIVAP SOLV SYS 230V 60HZ
Supplier: Labconco
The CentriVap Vacuum Concentration System^ Includes the following components: Benchtop Vacuum Concentrator with Glass Lid 7810014; -84 Cold Trap 7460020; Clear Canister 7460900; Solvent Insert 7815200; 12-13 mm Rotor 7455100; and Vacuum Tubing and 6 Clamps. 230V 60Hz NA
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CENTRIVAP SOLV SYS 230V 60HZ 89412-198 7995812 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty