Would you like to log in again?
KIT FIELD POWER FUMING SYS 115V PORB 6FT
Supplier: Labconco
Field Power Kit^ For CApture Portable Fuming System. Includes small bag, 2 batteries, charger, inverter and instructions. Allows CApture to run in remote locations for at least an hour on one battery. Once activated, batteries hold charge for one year. 115V
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT FIELD POWER FUMING SYS 115V PORB 6FT 89412-204 3426500 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty