Would you like to log in again?
PWRKT W/O BATTERIES 115V
Supplier: Labconco
Field Power Kit For Capture Portable Fuming System^ Includes Small Bag, Charger, Inverter And Instructions. Turnigy 5000Mah 3S 20C Lipo Pack Batteries Are Required And Sold Separately. Allows Capture To Run In Remote Locations For At Least An Hour On One Battery. Ea. 115V
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PWRKT W/O BATTERIES 115V 89428-596 3426501 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty