Would you like to log in again?
HEPA FILTR FOR PUMP/FREEZE DRY
Supplier: Labconco
Hepa Filter For All Labconco Freeze Dryers^ When Connecting To Labconco Rotary Vane Or Hybrid Vacuum Pumps. Filter Fits Between The Freeze Dryer And Pump To Prevent Particulate Contaminants From Getting Into The Pump. Ea.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HEPA FILTR FOR PUMP/FREEZE DRY 89428-600 7386500 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty