Would you like to log in again?
FreeZone® Freeze Dry Glassware Kit, Labconco
Supplier: Labconco
7540100
89428-604EA 794.71 USD InStock
89428-604
FreeZone® Freeze Dry Glassware Kit, Labconco
Freeze Dryers
Replacement glassware kit for FreeZone® freeze dry systems.
Delivery information: Includes one each 600 ml Complete Fast-Freeze Flask, one each 300 ml Complete Fast-Freeze Flask, one each 3/4" SST Straight Adapter, and one each 3/4" SST 45° Bend Adapter.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty