Would you like to log in again?
WaterPro® PS Polishing System, Labconco
Supplier: Labconco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WaterPro®
9000531 9000521 9047101 9000541 9000520 9000530 9000580 9000540 9000560 9000581 9000570 9000571 9000561
89496-482EA 6640 USD InStock
89496-482 89496-492 89496-484 89496-494 89496-486 89496-496 89496-488 89496-498 89496-500 89496-502 30181-416 89496-480 89496-490
WaterPro® PS Polishing System, Labconco
Water Purification Systems Deionizers
WaterPro PS Polishing Stations deliver Type I, 18.2 megohm-cm water from the dispensing valve or optional dispensing gun (refer to descriptions for models that include gun) at a typical rate of 1.8 liters per minute depending on feedwater flow rate and pressure, and installed purification modules.
The control panel includes a timed dispense feature that allows unattended dispensing of pure water without risk of overfilling. The dispenser shuts off automatically once user-set time has elapsed. 
Ordering information: Polishing kit (30181-416) is also required for operation and is sold separately. For available accessories, contact your Avantor representative.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty