Would you like to log in again?
WaterPro® PS/HPLC Analytical Instrument Model, Labconco
Supplier: Labconco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9000620 9000640 9000681 9000631 9000671 9000661 9000660 9000670 9000630 9000680 9000621 9000641
89496-508EA 7440 USD InStock
89496-508 89496-510 89496-520 89496-512 89496-522 89496-514 89496-524 89496-516 89496-504 89496-526 89496-518 89496-506
WaterPro® PS/HPLC Analytical Instrument Model, Labconco
Water Purification Systems Deionizers
WaterPro PS Polishing Stations deliver Type I, 18.2 megohm-cm water from the dispensing valve or optional dispensing gun (refer to descriptions for models that include gun) at a typical rate of 1.8 liters per minute depending on feedwater flow rate and pressure, and installed purification modules.
The control panel includes a timed dispense feature that allows unattended dispensing of pure water without risk of overfilling. The dispenser shuts off automatically once user-set time has elapsed. Labconco has designed WaterPro PS Stations with the right combinations of purification technologies to meet your needs.
The WaterPro PS/HPLC Analytical Instrument includes four bowls constructed of polypropylene for one carbon, two deionization cartridges and one organic adsorbtion cartridge (all cartridges sold separately).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty