Would you like to log in again?
WaterPro® PS HPLC/UF Hybrid, Labconco
Supplier: Labconco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9000722 9000762 9000772 9000732 9000742 9000782
89496-560EA 11970 USD InStock
89496-560 89496-552 89496-562 89496-554 89496-556 89496-558
WaterPro® PS HPLC/UF Hybrid, Labconco
Water Purification Systems Deionizers
WaterPro PS Polishing Stations deliver Type I, 18.2 megohm-cm water from the dispensing valve or optional dispensing gun (refer to descriptions for models that include gun) at a typical rate of 1.8 liters per minute depending on feedwater flow rate and pressure, and installed purification modules.
The control panel includes a timed dispense feature that allows unattended dispensing of pure water without risk of overfilling. The dispenser shuts off automatically once user-set time has elapsed. Labconco has designed WaterPro PS Stations with the right combinations of purification technologies to meet your needs.
The WaterPro PS/UF Hybrid model includes four bowls constructed of polypropylene for one carbon, two deionization and one organic adsorbtion cartridge (all cartridges sold separately).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty