Would you like to log in again?
ELECT RECEPT KIT 230V 13A 50HZ UK
Supplier: Labconco
Electrical Receptacle Kit^ For 3', 4', 5', 6', 7' Or 8' Protector Premier, Xstream, Xl, Pvc Perchloric Acid Or Stainless Steel Laboratory Hood. Includes Electrical Receptacle, Wiring, Junction Box And Receptacle Cover Plate. Ea. 230V, 13A, 50Hz Uk
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ELECT RECEPT KIT 230V 13A 50HZ UK 89501-526 9412500 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty