Would you like to log in again?
Protector® Stainless Steel Perchloric Acid Laboratory Hoods, Labconco®
Supplier: Labconco
130500020 130500002 130810001 130610001 130410020 130810002 130810020 130810021 130600020 130510020 130600000 130500021 130510000 130400002 130600021 130410001 130800020 130400021 130400020 130510021 130600001 130500000 130800002 130800001 130610002 130800000 130410002 130610020 130510002 130400001 130510001 130410000 130410021 130400000 130610000 130800021 130600002 130810000 130610021 130500001
89523-344EA 37060 USD InStock
89523-344 89523-322 89523-300 89523-288 89523-320 89523-342 89523-286 89523-304 89523-326 89523-346 89523-324 89523-302 89523-268 89523-280 89523-340 89523-284 89523-282 89523-318 89523-316 89523-338 89523-278 89523-310 89523-332 89523-298 89523-276 89523-330 89523-314 89523-336 89523-334 89523-312 89523-292 89523-270 89523-290 89523-296 89523-274 89523-294 89523-272 89523-308 89523-306 89523-328
Protector® Stainless Steel Perchloric Acid Laboratory Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Interior is one piece, Type 316 stainless steel with integral work surface and drainage trough.
Equipped with vapor-tight T8 fluorescent light fixture and light and blower switches. Two remote-controlled service fixtures can be used with gas, air, vacuum, or cold water. Each fixture has a serrated-tip hose connector. Hoods are factory prepared for up to eight service fixtures and four electrical duplexes. Hand-operated hood and duct washdown systems. Equipped with 27.9 cm (11") O.D., Type 316 stainless steel collar for connection to the exhaust stack.
Supporting work surface is included. Exterior is glacier white powder-coated steel with vertical, counter-balanced, tempered safety-glass sash.
Certifications: ETL and ETL-C listed. Meets NFPA (National Fire Protection Association) Standard 45, 'Fire Protection for Laboratories using Chemicals'.
Ordering information: The 4 to 6' models are available with 2 service fixtures and 1 electrical receptacle included. The 8' models are available with 2 service fixtures and 2 electrical receptacles included. For other accessories, contact your Avantor sales representative. A dedicated, remotely-located exhaust blower and a base cabinet or stand are required for operation (all sold separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty