Would you like to log in again?
XPert™ Bulk Powder Enclosures, Labconco®
Supplier: Labconco
XPert™
3963822 3963620 3963602 3963621 3963520 3963500 3963802 3963521 3963601 3963820 3963501 3963821 3963800 3963600 3963522 3963622 3963801 3963502
93000-408EA 43695 USD InStock
93000-408 93000-406 93000-404 93000-382 93000-394 93000-380 93000-392 93000-390 93000-402 93000-388 93000-400 93000-378 93000-386 93000-376 93000-398 93000-384 93000-374 93000-396
XPert™ Bulk Powder Enclosures, Labconco®
Fume Hoods and Enclosures Balances Enclosures
Bulk powder enclosures keep airborne particulates and fumes contained during powder transfer operations
Enclosures also feature a built-in Guardian™ 1000 digital airflow monitor. If the face velocity falls below the setpoint for more than six seconds, an audible alarm sounds and a red light glows. The alarm can be muted. Airflow monitor features an LCD display of actual face velocity and an RS-232 port. Retaining rings are constructed of Type 304 stainless steel. One retaining ring is required; it should be slightly larger than diameter of source container and a minimum of 5.1cm (2") smaller than the inner bag diameter. Polyethylene safety sleeves are fastened with a retaining band to the outside of the source container and access opening to help contain powders for easy clean up. The access opening cover provides a solid work surface. An extra transfer caddy is available to provide for easier material handling. Optional utility shelf kit has three epoxy-coated steel shelves.
Airfoils with Clean-Sweep™ openings enhance airflow, and a zone-perforated rear baffle creates a horizontal laminar airflow to maximize containment. A 30.5cm (12") sash operating height and the ample interior height can accommodate large micro or analytical balances. Angled sash is constructed of 0.6cm (1/4") thick tempered safety glass that is hinged to pivot up and lock for loading. Enclosures feature two utility ports, upper dilution air supplies, and upper containment sash foils. Enclosures incorporate a low-profile design and are equipped with side-entry airfoils, built-in top-mounted airflow monitor, and left side-mounted waste chute. Sides and top are constructed of 0.6cm (1/4") thick, non-static producing tempered safety glass that provides excellent visibility, ambient light, and protection. Sturdy, dry-powder and epoxy-coated aluminum frame and steel rear panel and baffle support the enclosure. A push-button hydraulic lift mechanism raises the container to the optimal height. An electronic beam senses the location of the container and triggers the lift mechanism to automatically stop at the optimal access height. Three 5.1x25.4cm (2x10") rectangular openings may be used to attach the enclosure to a remote blower or portable exhauster.
Certifications: All stations conform to modified ASHRAE 110-1995 standards, SEFA 1-2002 standards, and ANSI Z9.5-1-993 standards. The 115V models are ETL listed; the 230V models are CE marked.
Ordering information: Items designated as REDISHIP can be shipped within 48 hours of receiving your order. Enclosures are supplied with a transfer caddy with 7.6cm (3") casters. All enclosures require ducting to house exhaust, remote exhaust, or FilterMate™ portable exhausters (see 82010-632 series). All models also require a retaining ring, safety sleeves, retaining bands, access opening cover, and waste chute bags (all sold separately). Contact your VWR representative for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty