Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
XPert™ Filtered Balance Systems, Labconco®
Supplier: Labconco
XPert™
3950421 3950541 3950221 3950200 3950520 3950621 3950500 3950531 3950521 3950321 3950600 3950532 3950542 3950300 3950622 3950222 3950202 3950620 3950640 3950220 3950630 3950401 3950602 3950501 3950301 3950540 3950320 3950402 3950201 3950502 3950632 3950631 3950601 3950641 3950302 3950642 3950522 3950420 3950422 3950400 3950322 3950530
82018-114EA 10405 USD InStock
82018-114 93000-518 82018-116 93000-516 93000-538 82018-110 93000-536 82018-112 93000-558 82018-118 89004-482 89004-484 93000-522 93000-544 89004-486 82018-120 93000-520 93000-542 89004-488 93000-540 93000-562 93000-560 82018-126 82018-128 93000-528 93000-548 93000-526 82018-122 93000-524 93000-546 82018-124 82018-108 89004-490 89004-492 93000-534 93000-556 93000-532 93000-554 82018-130 93000-530 93000-552 93000-550
XPert™ Filtered Balance Systems, Labconco®
Fume Hoods and Enclosures Balances Enclosures
These portable systems provide user protection by keeping powders and particulates contained during weighing procedures
The deep interior easily accommodates large micro and analytical balances. The Clean-Sweep™ airfoil allows air to sweep the work surface for maximum containment. The perforated rear baffle promotes horizontal laminar airflow to maximize containment. The upper containment sash foil bleeds air into the enclosure to direct concentrations of contaminants away from the operator’s breathing zone. The upper dilution air supply introduces room air at the top of the sash to dilute contaminants in the upper chamber and bathe the back of the sash with clean air. Additional side-entry airfoils allow air to sweep across the interior glass surfaces to enhance containment. The airfoil’s ergonomic design provides a comfortable position for the operator’s forearms. Systems feature a true bag-in/bag-out filter disposal system for safely removing and replacing the HEPA filter. The HEPA filter cell is 99.99% efficient on particulates 0.3µm in size. An upper diffuser screen, located above the work area, provides protection for the HEPA filter. The system also features a front-mounted Minihelic® pressure gauge that monitors pressure differential across the HEPA filter to help determine filter loading. The built-in, positive pressure impeller is surrounded by a negative pressure plenum so that if a leak should develop, the unfiltered air is captured and refiltered. The blower also has vibration isolation supports to minimize disturbance to sensitive balances.
Sturdy glacier white and gray, dry powder epoxy-coated aluminum frame and steel rear plenum and baffle dissipate static charge. Tempered safety glass front sash, sides, and top offer excellent visibility, ambient light, and protection. Ergonomic angled sash allows a closer view, reduces glare, and provides a more comfortable operating position than vertical sashes. Hinged sash pivots upward and locks to a loading height of 48.7cm (19"). The 1.5m (5') and 1.8m (6') models incorporate a gas-assist lift and have a loading height of 70.4cm (2711/16"). The operating sash height is 23.9cm (913/32") on the 0.6m (2'), 0.9m (3'), and 1.2m (4') models. The 1.5m (5') and 1.8m (6') models have a 30.5cm (12") operating sash height. A wiping seal helps keep contaminants contained. Systems also include front-mounted light and blower switches, two utility ports, and glare-free fluorescent lighting.
Enclosures also feature an optional built-in airflow monitor. If the hood’s face velocity falls below the setpoint for more than six seconds, an audible alarm sounds and a red light glows. The alarm can be muted. On the Guardian™ monitor, an LED glows green when the airflow is within setpoint range. The Guardian™ 1000 digital monitor features an LCD display of actual face velocity and an RS-232 port. Optional trace odor carbon filters can be easily installed atop the system to trap nuisance vapors from organic solvents, formaldehyde, ammonia, or amines. Stainless steel work surfaces feature a spill trough. The utility shelf kit has three epoxy-coated steel shelves that can hold a printer, utensils, and other miscellaneous supplies. Shelves attach to the rear baffle perforations of the enclosure.
All systems conform to modified ASHRAE 110-1995 standards, SEFA 1-2002 standards, and ANSI Z9.5-1-993 standards. The 115V systems are ETL and ETL-C listed; the 230V systems are CE marked.
The built-in blower and HEPA filter are mounted in the upper plenum to save space, and the enclosures have low installation costs and require no outside ducting. Systems also provide quiet operation as low as 45dBA, depending on face velocity.
Ordering information: Items designated below as REDISHIP can be shipped within 48 hours of receiving your order. All systems require a work surface and supporting base. For base cabinets, see the Protector® acid storage cabinets (46610-412 series), solvent storage cabinets (46610-440 series), and standard cabinets (46610-404 series). The 1.5m (5') and 1.8m (6') models with 90.2cm (351/2") exterior depth include a left side-mounted waste chute that accepts bags 25.4x40.6cm (10x16") and larger. The 230V models do not include a plug. Trace odor carbon filters are also available; contact your VWR® sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty