Would you like to log in again?
Base Stands and Accessories for Purifier® Logic® Series Class II Safety Cabinets, Labconco®
Supplier: Labconco
3780310 3859601 3780106 3858502 3780100 3859502 3780203 3811105 3780104 3780103 3780200 3288900 3784000 8022000 3288902 3730410 3859603 3858800 3780202 3780201 3858601 38305003726 3830510 3811107 3288901 3780311 3859400P 3858500 3780107 3780105 3869300 37306003726 37303003726 3858603 3858501 3859000 3859501 3850200 3811106 3780313 3859600 3780101 3730610 3730310 3869200 3288903 3780312 3859602 3858600 3858602 3859100 3811108 3857000 3859500 3859503 7071900 3747500 3780102 3852000 3850100 3730410
89003-096EA 4655 USD InStock
89003-096 82003-608 97006-960 89003-094 82003-606 97010-022 97007-004 97007-006 97006-958 97006-956 89003-092 97006-954 89003-090 97006-952 83009-284 97006-950 97007-016 97007-018 46610-764 97006-428 97006-426 97006-986 97006-424 97006-984 97006-422 82013-198 82013-194 82003-588 82013-190 97007-028 77574-600 82003-592 97007-034 97007-594 82003-590 97007-036 97007-596 77574-604 97007-030 470203-158 82003-630 77574-602 97007-032 43100-012 82013-202 89003-088 82003-578 82003-612 82003-576 77574-606 82003-610 97007-038 82003-616 43100-014 82003-614 97007-000 82003-580 97007-002 97007-040 97007-042 97006-966
Base Stands and Accessories for Purifier® Logic® Series Class II Safety Cabinets, Labconco®
Fume Hoods and Enclosures PCR Workstation Enclosures
Logic Cart fits under any telescoping base stand. Features three small and one large compartments, one drawer, front panel cut out for dispensing wipers or gloves, and HDPE worksurface. W×D×H: 635×483×686 mm (25×19×27").
Service Fixture Kit includes serrated hose tip valve with ball valve, quarter turn control handle, hardware and instructions for plumbing to services. Mounts on left or right side interior. Up to two may be installed per side.
Base Stands — Constructed of durable 45 mm (1¾") tubular, glacier white epoxy-coated steel, these ADA-compliant base stands support up to 453.6 kg (1000 lbs.). Stands have a depth of 737 mm (29"). Stands with telescoping legs are adjustable to seven height positions in 25 mm (1") increments from 699 to 851 mm (27½ to 33½"). Available with fixed leveling feet or 127 mm (5") diameter toe-locking polyurethane casters with bearings for quiet operation. Hydraulic lift base stands are adjustable to any height from 648 to 851 mm (25½ to 33½") and include four powder-coated, die-cast feet with levelers. Feet can be replaced with casters for mobility. Manual hydraulic lift models include front-accessible hand crank. Electric hydraulic lift models include a front-accessible up/down switch and a 3.7 m (12') three-wire cord and plug. Electrical components of electric hydraulic lift stands are UL listed.
They support a maximum of 589.7 kg (1300 lbs.). Stands have a depth of 870 mm (34⅟₄"). Built-in hydraulic lift is operated by remote control with buttons for upward and downward adjustability, providing work surface heights from 43.8 to 93.3 cm (17¼ to 36¾") and overall enclosure height as low as 1935 mm (76.2"). The supported enclosure can be positioned for persons of different statures, wheelchair-bound users, transport through doorways, and easier access to upper components. Stands include 127 mm (5") diameter toe-locking polyurethane casters with bearings for quiet operation. Handle bars on both sides facilitate transport. When placed on stands that can be easily raised and lowered, enclosures that are ducted outside will require special modifications to incorporate flexible ducting, wiring, and plumbing. Placing outside ducted enclosures on mobile stands is not recommended.
Caster Kit contains four 127 mm (5") diameter casters. For use with hydraulic lift base stands, except for the SoLo base stands, which come with casters. Shelves are epoxy-coated 25 cm (9²⁹/₃₂") deep steel shelves for use with telescoping base stands.
Certifications: SoLo™ Base Stands with Remote Control are ADA- and NSF-compliant.
Ordering information: All base stands for 1.2 m (4') and 1.8 m (6') models can also be used with PuriCare® Procedure Stations.
Delivery information: Items designated below as REDISHIP can be shipped within 48 hours of receiving your order.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty