Would you like to log in again?
FLTR FLASH NYL 25MM .2U PK100
Supplier: Agela Technologies
Inline Flash Cartridge filter^ To safeguard detectors and fraction collectors ; Compatible with methanol, acetonitrile and aqueous solvents; Housing: high purity PP; Membrane: Nylon, 0.22 um pore size; 25 mm in diameter
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLTR FLASH NYL 25MM .2U PK100 97011-450 AS022520-P Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty