Would you like to log in again?
Mobile Equipment Table, Labconco®
Supplier: Labconco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3819100 3819101 3819102
97025-420EA 2350 USD InStock
97025-420 97025-422 97025-418
Mobile Equipment Table, Labconco®
Furniture Tables and Workbenches
Mobile equipment table may be used to transport heavy equipment and to provide auxiliary bench space.
  • 5" diameter non-marking polyurethane casters with toe locks have bearings for quiet operation
  • Supports loads up to 800 pounds
With height adjustments from 28.75" to 35.75", it may be raised to the same height as most laboratory countertops. Solid epoxy 1.25" thick × 30" deep work surface and epoxy-coated 1.75" square tubular steel frame.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty