Would you like to log in again?
XPert™ Nano Ventilated Enclosures, Labconco®
Supplier: Labconco
XPert™
3888621 3888662 3887560 3887220 3887260 3887362 3887221 3887660 3887520 3887662 3887361 3888521 3887262 3887460 3887421 3887420 3888520 3887321 3888561 3887562 3888620 3887320 3887620 3887661 3888661 3887561 3887621 3887462 3888562 3887461 3888660 3888560 3887261 3887521 3887360
97025-454EA 18970 USD InStock
97025-454 97025-476 97025-432 97025-474 97025-452 97025-430 97025-458 97025-436 97025-434 97025-478 97025-456 97025-490 97025-472 97025-450 97025-492 97025-470 97025-428 97025-426 97025-448 97025-486 97025-442 97025-464 97025-440 97025-462 97025-484 97025-424 97025-468 97025-446 97025-444 97025-466 97025-488 97025-460 97025-482 97025-480 97025-438
XPert™ Nano Ventilated Enclosures, Labconco®
Fume Hoods and Enclosures Extraction Cabinets
During procedures such as nanoparticle manipulation and dry powder chemical handling, enclosures keep hazardous powders and particulates contained
Features deep interiors, Guardian™ airflow monitor, true bag-in/bag-out filter disposal system, built-in blower, ergonomic air foil with aerodynamic Clean-Sweep™ openings that enhance airflow, upper dilution air supply, upper containment sash foil, zone-perforated rear baffle, and pressure gauge for monitoring filter loading. Type 304 stainless steel interior sides, removable baffle, removable air foil, and integral work surface are easy to wipe down and clean. Exterior is constructed of glacier white and gray dry powder epoxy-coated aluminum and steel.
Optional built-in ionizer neutralizes static charge on interior surfaces by emitting ions into the airstream, which reduces weighing errors and the attraction of powders to surfaces.
The 115V units are ETL and ETL-C listed; 230V models are CE marked.
Room air is pulled into the enclosure from the front, flows to the baffle, and finally passes through a ULPA exhaust filter (99.999% efficient on particles 0.12 micron in size) before returing to the laboratory or cleanroom. ULPA filter is top-accessible for leak scanning of entire surface.
Ordering information: All models require a supporting base (not included). Optional accessories include a canopy connection, remote blowers, and ductwork. Contact your Avantor representative for more information. Bag-in/bag-out filter disposal system feature requires a certified technician.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty