Would you like to log in again?
Mini Stoppering Chamber for FreeZone® Freeze Dryers, Labconco
Supplier: Labconco
7569000
89428-602EA 2047.31 USD InStock
89428-602
Mini Stoppering Chamber for FreeZone® Freeze Dryers, Labconco
Freeze Dryers
The Mini Stoppering Chamber is ideal for lyophilizing samples in small serum vials with split stoppers.
Simply attach the Mini Stoppering Chamber to a port on a FreeZone Freeze Dry System (sold separately). Once the samples are lyophilized, the vials can be stoppered while still under vacuum. A tray is included to easy loading and unloading of samples. Can accommodate up to forty-eight 2 mL serum vials or eight 30 mL serum vials and has a stoppering travel distance of .5". Can accommodate vials that range in height from 1.38" to 2.48" tall.
Ordering information: Vials and stoppers are sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty