Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Imiquimod 99%

Supplier: SELLECK CHEMICALS LLC MS
Synonyms: 1-Isobutyl-1H-imidazo[4,5-c]quinolin-4-amine4-Amino-1-isobutyl-1H-imidazo[4,5-c]quinoline

S1211-100MG S1211-200MG
101759-614EA 107 USD InStock
101759-614 101759-612
Imiquimod 99%
Imiquimod
Imiquimod is a novel synthetic agent with immune response modifying activity.

  • Powder
  • Solubility: 1 mg/mL warmed (DMSO)
  • Insoluble in water


Formula: C₁₄H₁₆N₄
MW: 240.31 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00866946
CAS Number: 99011-02-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty