Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ciclesonide 99.32%

Supplier: SELLECK CHEMICALS LLC MS
Warning
Synonyms: OmnarisCiclesonide

S4646-20MG S4646-100MG
103544-872EA 192.31 USD InStock
103544-872 103544-874
Ciclesonide 99.32%
Ciclesonide
Ciclesonide is a glucocorticoid used to treat obstructive airway diseases.


Formula: C₃₂H₄₄O₇
MW: 540.69 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00866004
CAS Number: 126544-47-6
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Purity (HPLC) 99.32%
NMR Consistent with structure

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty