Would you like to log in again?
FreeZone® Support Shelves, Labconco®
Supplier: Labconco
FreeZone®
7426400
82010-516PK 186.31 USD InStock
82010-516
FreeZone® Support Shelves, Labconco®
Furniture Shelves
Attach easily without tools to the sides of the FreeZone cabinet.
  • It provides support for large flasks attached to the drying chamber or manifold
  • Glacier white epoxy-coated steel wire
Multiple rows of perforations on the sides of the FreeZone cabinet allow for height adjustment of the shelves.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty