Would you like to log in again?
Colorsmart Filter for pre-2005 FilterMate Portable Exhausters, Labconco®
Supplier: Labconco
6933201 6910501 6910401 6910301 6910201
46000-244EA 1455 USD InStock
46000-244 46000-242 46000-236 46000-238 46000-240
Colorsmart Filter for pre-2005 FilterMate Portable Exhausters, Labconco®
Filters Gas Filters
Carbon filter for pre-2005 FilterMate portable exhausters.
Contain a minimum of 26 pounds of impregnated carbon. Color-coded. Includes three sampling tubes that detect a broad range of chemicals specific to the filter. For use as replacement filter in discontinued Protector FilterMate Mobile Stations currently in service.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty